missing translation for 'onlineSavingsMsg'
Learn More

Methylaminoacetonitrile hydrochloride, 98%

Catalog No. p-7036807
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5521506 5 g
AAH5521514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5521506 Supplier Thermo Scientific Chemicals Supplier No. H5521506
Only null left

CAS: 25808-30-4 | C3H7ClN2 | 106.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25808-30-4
Molecular Formula C3H7ClN2
Molecular Weight (g/mol) 106.553
MDL Number MFCD00012587
InChI Key DFKBQHBEROHUNF-UHFFFAOYSA-N
Synonym methylaminoacetonitrile hydrochloride, 2-methylamino acetonitrile hydrochloride, acetonitrile, methylamino-, monohydrochloride, sarcosinenitrile hydrochloride, n-methylaminoacetonitrile hydrochloride, acetonitrile, 2-methylamino-, hydrochloride 1:1, methylamino acetonitrilhydrochlorid, methylaminoacetonitrile, hydrochloride, methylaminoacetonitrilehydrochloride, cyanomethyl methyl ammonium chloride
PubChem CID 117641
IUPAC Name 2-(methylamino)acetonitrile;hydrochloride
SMILES CNCC#N.Cl

Specifications

Melting Point 103°C to 106°C
Linear Formula CH3NHCH2CN·HCl
Quantity 5 g
Beilstein 3908975
Formula Weight 106.55
Percent Purity 99%
Chemical Name or Material Methylaminoacetonitrile hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 247-277-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only