missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet, Reagent Grade, LabChem™

Catalog No. LC171907
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC171907 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC171907 Supplier LabChem Supplier No. LC171907
Only null left

Chemical Identifiers

CAS 8004-87-3

Specifications

Melting Point 137°C
CAS Max % 100
Linear Formula C24H28N3Cl
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 393.96
Grade Reagent
Packaging Amber Glass
Decomposition Information Carbon monoxide; Carbon dioxide
Color Green
Quantity 10 g
Physical Form Powder
Chemical Name or Material Methyl Violet
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature