missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet Indicator, 0.02% (w/v), Ricca Chemical

$78.57 - $78.57

Chemical Identifiers

CAS 8004-87-3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5100-4
View Documents
Ricca Chemical Company
51004
120 mL Natural Poly Bottle
Each for $78.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

8004-87-3

Specifications

8004-87-3
Indicator
Natural Poly Bottle
>1.6
Methyl Violet
0.02% W/V
0.02% (w/v) Aqueous Solution
Purple
120 mL
Videos
SDS
Documents

Documents