missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Violet 2B, pure, indicator grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 137.0°C |
| Absorption Ratio | 1 to 1.25 |
| Infrared Spectrum | Authentic |
| Lambda Max | 580 to 586nm (in 50:50 IMS:water) |
| Loss on Drying | 10% max. (105°C) |
| Packaging | Glass bottle |
| UV_Vis | 75% min. (on dry substance) |
| Beilstein | 13, 755 |
| Color | Green |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| RTECSNumber | GC8425000 |
| TSCA | TSCA |
RUO – Research Use Only