missing translation for 'onlineSavingsMsg'
Learn More

Methyl Violet 2B, pure, certified

CAS: 8004-87-3 | C24H28ClN3

Supplier:  Thermo Scientific Chemicals 415070250

 View more versions of this product

Catalog No. AC415070250

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl Violet 2B
Pure
1.06 to 1.19
C24H28ClN3
13, 755
Solubility in water: soluble. Other solubilities: soluble in conc. sulfuric acid and nitric acid, soluble in chloroform, partially soluble in alcohol and glycerol
583 to 587nm
Pure
Green
Crystalline Powder With Metal Luster
137.0°C
8004-87-3
75% min.
MFCD00012568
Basic Violet 1, C.I. 42535
Authentic
393.95
Glass bottle
25 g

Chemical Identifiers

8004-87-3
C24H28ClN3
MFCD00012568
Basic Violet 1, C.I. 42535
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only