missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Violet 2B

Catalog No. AAA1215918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1215918 50 g
AAA1215930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1215918 Supplier Thermo Scientific Chemicals Supplier No. A1215918

CAS: 8004-87-3 | C24H27N3·HCl

Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton?s crystal violet-iodine technique for chromatin and nucleoli; Lieb?s amyloid stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.

Solubility
Soluble in water.

Notes
Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H27N3·HCl
MDL Number MFCD00012568
Synonym Basic Violet 1; C.I. 42535

Specifications

Solubility Information Soluble in water.
Formula Weight 393.96
Odor Odorless
Quantity 50 g
Chemical Name or Material Methyl Violet 2B
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
RTECSNumber GC8425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only