missing translation for 'onlineSavingsMsg'
Learn More

Methyl vinyl sulfone, 95%, stabilized, Thermo Scientific™

Chemical Identifiers

CAS 3680-02-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00007567
InChI Key WUIJTQZXUURFQU-UHFFFAOYSA-N
Synonym zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl
PubChem CID 100016
ChEBI CHEBI:46938
IUPAC Name methanesulfonylethene
SMILES CS(=O)(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306730010
SDS View Documents
Thermo Scientific Chemicals
306730010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC306730500
SDS View Documents
Thermo Scientific Chemicals
306730500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3680-02-2
106.14
WUIJTQZXUURFQU-UHFFFAOYSA-N
100016
methanesulfonylethene
C3H6O2S
MFCD00007567
zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl
CHEBI:46938
CS(=O)(=O)C=C

Specifications

3680-02-2
115.0°C to 117.0°C (19.0 mmHg)
Authentic
Glass bottle
1.4610 to 1.4630
MFCD00007567
1.21
WUIJTQZXUURFQU-UHFFFAOYSA-N
1-methylsulfonylethene
150ppm 4-tert.-butylpenol
106.14
Methyl vinyl sulfone, Stabilized
1.2100g/mL
>110°C
94% min. (GC)
C3H6O2S
H2C=CHSO2CH3
1g
methyl vinyl sulfone, ethene, methylsulfonyl, methyl vinyl sulphone, methylvinylsulfone, methanesulfonylethene, sulfone, methyl vinyl, vinyl methyl sulfone, methanesulfonyl-ethene, methylsulfonyl ethene, methyl vinyl sulfphone
CS(=O)(=O)C=C
106.14
19354
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Fatal in contact with skin.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-958-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only