missing translation for 'onlineSavingsMsg'
Learn More

Methyl vinyl ketone, tech. 90%, stab.

Catalog No. p-7081769
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11910AC 25 mL
AAA11910AE 100 mL
AAA11910AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA11910AC Supplier Thermo Scientific Chemicals Supplier No. A11910AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-94-4 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-94-4
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00008777
InChI Key FUSUHKVFWTUUBE-UHFFFAOYSA-N
Synonym methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
PubChem CID 6570
ChEBI CHEBI:48058
IUPAC Name but-3-en-2-one
SMILES CC(=O)C=C

Specifications

Melting Point -7°C
Density 0.856
Boiling Point 32°C to 34°C (60 mmHg)
Flash Point −6°C (21°F)
Assay Percent Range 90%, Stabilized with 0.5% Hydroquinone
Refractive Index 1.412
Quantity 25 mL
UN Number UN1251
Beilstein 506021
Merck Index 14,6134
Formula Weight 70.09
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl vinyl ketone, Stabilized
Show More Show Less
Hazard Category H225-H300+H310+H330-H314-H317-H335
Hazard Statement GHS H Statement
H224-H300-H330-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL VINYL KETONE, STABILIZED
EINECSNumber 201-160-6
RTECSNumber EM9800000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only