missing translation for 'onlineSavingsMsg'
Learn More

Methyl valerate, 99%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 415050250

Catalog No. AC415050250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl valerate
624-24-8
0.8900g/mL
29°C
98.5% min. (GC)
C6H12O2
CH3(CH2)3COOCH3
25g
0.89
Solubility in water: insoluble
CCCCC(=O)OC
116.16
116.16
99%
1mg KOH/g max.
126°C
Authentic
Glass bottle
1.3965 to 1.3985
MFCD00009478
02, 301
methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
HNBDRPTVWVGKBR-UHFFFAOYSA-N
methyl pentanoate
12206
99%

Chemical Identifiers

624-24-8
116.16
HNBDRPTVWVGKBR-UHFFFAOYSA-N
12206
CCCCC(=O)OC
C6H12O2
MFCD00009478
methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
methyl pentanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.

GHS Signal Word: Warning

EINECSNumber : 210-838-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only