missing translation for 'onlineSavingsMsg'
Learn More

Methyl undecanoate, 99%

Catalog No. AAL0593906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0593906 5 g
AAL0593914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0593906 Supplier Thermo Scientific Chemicals Supplier No. L0593906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1731-86-8 | C12H24O2 | 200.322 g/mol

Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

Solubility
Soluble in ethanol, and ether. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1731-86-8
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00008957
InChI Key XPQPWPZFBULGKT-UHFFFAOYSA-N
Synonym undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
PubChem CID 15607
ChEBI CHEBI:87527
IUPAC Name methyl undecanoate
SMILES CCCCCCCCCCC(=O)OC

Specifications

Melting Point -10°C
Density 0.871
Boiling Point 246°C to 248°C
Flash Point 109°C (228°F)
Refractive Index 1.429
Quantity 5 g
Beilstein 1762129
Solubility Information Soluble in ethanol,and ether. Insoluble in water.
Percent Purity 99%
Chemical Name or Material Methyl undecanoate
Hazard Category H315-H318-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 217-053-2
RTECSNumber YQ2573000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only