missing translation for 'onlineSavingsMsg'
Learn More

Methyl undecanoate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013340
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013340 1 mL
111013341 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013340 Supplier Merck Emd Millipore Supplier No. 941181ML

Specifications

CAS 1731-86-8
Density 0.872 g/mL (at 25°C)
Flash Point 109°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)9 CO2 CH3
MDL Number MFCD00008957
Synonym Undecanoic acid methyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 200.32
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less