missing translation for 'onlineSavingsMsg'
Learn More

Methyl (trimethylsilyl)propiolate, 98%

CAS: 42201-71-8 | C7H12O2Si | 156.256 g/mol

$217.14 - $764.38

Chemical Identifiers

CAS 42201-71-8
Molecular Formula C7H12O2Si
Molecular Weight (g/mol) 156.256
MDL Number MFCD04039783
InChI Key XXGHAMBKHIDZDC-UHFFFAOYSA-N
Synonym methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #
PubChem CID 560731
IUPAC Name methyl 3-trimethylsilylprop-2-ynoate
SMILES COC(=O)C#C[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2524503
View Documents
Thermo Scientific Chemicals
B2524503
1 g
Each for $217.14
Only null left
 
AAB2524506
View Documents
Thermo Scientific Chemicals
B2524506
5 g
Each for $764.38
Only null left
 
Description

Description

Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Chemical Identifiers

42201-71-8
156.256
XXGHAMBKHIDZDC-UHFFFAOYSA-N
560731
COC(=O)C#C[Si](C)(C)C
C7H12O2Si
MFCD04039783
methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #
methyl 3-trimethylsilylprop-2-ynoate

Specifications

71°C to 75°C (11mmHg)
1 g
(CH3)3SiC≡CCO2CH3
UN3272
XXGHAMBKHIDZDC-UHFFFAOYSA-N
methyl 3-trimethylsilylprop-2-ynoate
560731
98%
Methyl (trimethylsilyl)propiolate
42201-71-8
C7H12O2Si
MFCD04039783
methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #
COC(=O)C#C[Si](C)(C)C
156.256
156.25
1.44
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only