accessibility menu, dialog, popup

UserName

CAS RN 42201-71-8

IUPAC Name: methyl 3-(trimethylsilyl)prop-2-ynoate
Synonyms: methyl 3-(trimethylsilyl)prop-2-ynoate
Molecular Weight (g/mol): 156.26
Molecular Formula: C7H12O2Si
InChi Key: XXGHAMBKHIDZDC-UHFFFAOYSA-N
SMILES: COC(=O)C#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results