missing translation for 'onlineSavingsMsg'
Learn More

Methyl Trifluoromethanesulfonate 98.0+%, TCI America™

Supplier:  TCI America T202925G

 View more versions of this product

Catalog No. T202925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Trifluoromethanesulfonate
Yellow
C2H3F3O3S
25 g
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
COS(=O)(=O)C(F)(F)F
164.10
164.10
Liquid
333-27-7
99°C
MFCD00000409
2920
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
methyl trifluoromethanesulfonate
9526
≥98.0% (GC)

Chemical Identifiers

333-27-7
164.10
OIRDBPQYVWXNSJ-UHFFFAOYSA-N
9526
COS(=O)(=O)C(F)(F)F
C2H3F3O3S
MFCD00000409
methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
methyl trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions