missing translation for 'onlineSavingsMsg'
Learn More

Methyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7137068
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20295G 5 g
T202925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20295G Supplier TCI America Supplier No. T20295G
Only null left

Chemical Identifiers

CAS 333-27-7
Molecular Formula C2H3F3O3S
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000409
InChI Key OIRDBPQYVWXNSJ-UHFFFAOYSA-N
Synonym methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
PubChem CID 9526
IUPAC Name methyl trifluoromethanesulfonate
SMILES COS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
UN Number 2920
Formula Weight 164.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Trifluoromethanesulfonate