missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tridecanoate 98.0+%, TCI America™

Supplier:  TCI America T096025ML

Catalog No. T096025ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Tridecanoate
7°C
163°C
MFCD00008977
tridecanoic acid, methyl ester, tridecanoic acid methyl ester, methyl n-tridecanoate, unii-o2h463ring, o2h463ring, tridecanoic acid,methyl ester, n-tridecanoic acid methyl ester, methyl tridecylate, methyltridecanoate, tridecanoic acid methyl
CCCCCCCCCCCCC(=O)OC
228.376
228.38
Liquid
1731-88-0
Yellow
C14H28O2
25 mL
JNDDPBOKWCBQSM-UHFFFAOYSA-N
methyl tridecanoate
15608
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : YD3950000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions