missing translation for 'onlineSavingsMsg'
Learn More

Methyl tricosanoate, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013668
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013668 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013668 Supplier Merck Emd Millipore Supplier No. 914781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2433-97-8
Physical Form Neat
Linear Formula CH3(CH2)21 COOCH3
MDL Number MFCD00009349
Melting Point 53°C to 56°C; 53°C to 56°C
Synonym Tricosanoic acid methyl ester
Quantity 1 g
Formula Weight 368.64
Percent Purity ≥99.0% (GC)
Grade Analytical Standard