missing translation for 'onlineSavingsMsg'
Learn More

Methyl tricosanoate, 99+%

Catalog No. p-4089644
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428450010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428450010 Supplier Thermo Scientific Chemicals Supplier No. 428450010
Only null left

CAS: 2433-97-8 | C24H48O2 | 368.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2433-97-8
Molecular Formula C24H48O2
Molecular Weight (g/mol) 368.65
MDL Number MFCD00009349
InChI Key VORKGRIRMPBCCZ-UHFFFAOYSA-N
Synonym tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
PubChem CID 75519
IUPAC Name methyl tricosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 53°C
Flash Point >113°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)21CO2CH3
Quantity 1 g
Formula Weight 368.64
Percent Purity 99+%
Chemical Name or Material Methyl tricosanoate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-420-2

RUO – Research Use Only