missing translation for 'onlineSavingsMsg'
Learn More

Methyl tricosanoate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013669
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013669 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013669 Supplier Merck Emd Millipore Supplier No. 91478250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2433-97-8
Flash Point >113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)21 COOCH3
MDL Number MFCD00009349
Melting Point 53°C to 56°C; 53°C to 56°C
Synonym Tricosanoic acid methyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 368.64
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less