missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Triacontanate 95.0+%, TCI America™
$308.03 - $1676.82
Chemical Identifiers
| CAS | 629-83-4 |
|---|---|
| Molecular Formula | C31H62O2 |
| Molecular Weight (g/mol) | 466.835 |
| MDL Number | MFCD00042897 |
| InChI Key | BIRUBGLRQLAEFF-UHFFFAOYSA-N |
| Synonym | methyl triacontanate, methyl melissate, melissic acid methyl ester, methyl melissicate, triacontanoic acid methyl ester, triacontanoic acid, methyl ester, n-triacontanoic acid methyl ester, triacontanoic acid methylester, methyltriacontanate, acmc-1b46i |
| PubChem CID | 12400 |
| IUPAC Name | methyl triacontanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
T0812100MG
|
TCI America
T0812100MG |
100 mg |
Each for $308.03
|
|
|||||
|
T08121G
|
TCI America
T08121G |
1 g |
Each for $1,676.82
|
|
|||||
Chemical Identifiers
| 629-83-4 | |
| 466.835 | |
| BIRUBGLRQLAEFF-UHFFFAOYSA-N | |
| 12400 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC |
| C31H62O2 | |
| MFCD00042897 | |
| methyl triacontanate, methyl melissate, melissic acid methyl ester, methyl melissicate, triacontanoic acid methyl ester, triacontanoic acid, methyl ester, n-triacontanoic acid methyl ester, triacontanoic acid methylester, methyltriacontanate, acmc-1b46i | |
| methyl triacontanoate |
Specifications
| 629-83-4 | |
| White | |
| MFCD00042897 | |
| methyl triacontanate, methyl melissate, melissic acid methyl ester, methyl melissicate, triacontanoic acid methyl ester, triacontanoic acid, methyl ester, n-triacontanoic acid methyl ester, triacontanoic acid methylester, methyltriacontanate, acmc-1b46i | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 466.835 | |
| 466.84 | |
| Crystalline Powder |
| 72°C | |
| C31H62O2 | |
| 100 mg | |
| BIRUBGLRQLAEFF-UHFFFAOYSA-N | |
| methyl triacontanoate | |
| 12400 | |
| ≥95.0% (GC) | |
| Methyl Triacontanate |
Safety and Handling
TSCA : No