missing translation for 'onlineSavingsMsg'
Learn More

Methyl Triacontanate 95.0+%, TCI America™

Catalog No. T0812100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0812100MG 100 mg
T08121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0812100MG Supplier TCI America Supplier No. T0812100MG

Chemical Identifiers

CAS 629-83-4
Molecular Formula C31H62O2
Molecular Weight (g/mol) 466.835
MDL Number MFCD00042897
InChI Key BIRUBGLRQLAEFF-UHFFFAOYSA-N
Synonym methyl triacontanate, methyl melissate, melissic acid methyl ester, methyl melissicate, triacontanoic acid methyl ester, triacontanoic acid, methyl ester, n-triacontanoic acid methyl ester, triacontanoic acid methylester, methyltriacontanate, acmc-1b46i
PubChem CID 12400
IUPAC Name methyl triacontanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 72°C
Color White
Quantity 100 mg
Formula Weight 466.84
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Triacontanate