missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-vaccenate, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001956
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001956 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001956 Supplier Merck Emd Millipore Supplier No. 0758625MG

Specifications

CAS 6198-58-9
Linear Formula C19H36O2
MDL Number MFCD00064484
Synonym trans-11-Octadecenoic acid methyl ester; Methyl trans-11-octadecenoate
Type Analytical Standard
Quantity 25 mg
Formula Weight 296.49
Percent Purity ≥95% (GC)
Grade Analytical Standard