missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-p-Coumarate 98.0+%, TCI America™

Catalog No. p-7133118
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22595G 5 g
M225925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22595G Supplier TCI America Supplier No. M22595G

Chemical Identifiers

CAS 19367-38-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00157167
InChI Key NITWSHWHQAQBAW-UHFFFAOYSA-N
Synonym trans-p-Coumaric Acid Methyl Ester, trans-4-Hydroxycinnamic Acid Methyl Ester, Methyl trans-4-Hydroxycinnamate
PubChem CID 5319562
ChEBI CHEBI:86904
IUPAC Name methyl 3-(4-hydroxyphenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(O)C=C1

Specifications

Melting Point 139°C
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl trans-p-Coumarate