accessibility menu, dialog, popup

UserName

CAS RN 19367-38-5

IUPAC Name: methyl 3-(4-hydroxyphenyl)prop-2-enoate
Synonyms: methyl hydroxycinnamate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: NITWSHWHQAQBAW-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results