missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl trans-9-Octadecenoate 98.0+%, TCI America™
$87.60 - $255.38
Chemical Identifiers
| CAS | 1937-62-8 |
|---|---|
| Molecular Formula | C19H36O2 |
| Molecular Weight (g/mol) | 296.495 |
| MDL Number | MFCD00066521 |
| InChI Key | QYDYPVFESGNLHU-ZHACJKMWSA-N |
| Synonym | methyl elaidate, elaidic acid methyl ester, methyl octadec-9-enoate, methyl trans-9-octadecenoate, e-methyl octadec-9-enoate, methyl 9-octadecenoate, 9-octadecenoic acid, methyl ester, 9-octadecenoic acid, methyl ester, e, 9-octadecenoic acid, methyl ester, 9e, methyl e-octadec-9-enoate |
| PubChem CID | 5280590 |
| IUPAC Name | methyl (E)-octadec-9-enoate |
| SMILES | CCCCCCCCC=CCCCCCCCC(=O)OC |
Chemical Identifiers
| 1937-62-8 | |
| 296.495 | |
| QYDYPVFESGNLHU-ZHACJKMWSA-N | |
| 5280590 | |
| CCCCCCCCC=CCCCCCCCC(=O)OC |
| C19H36O2 | |
| MFCD00066521 | |
| methyl elaidate, elaidic acid methyl ester, methyl octadec-9-enoate, methyl trans-9-octadecenoate, e-methyl octadec-9-enoate, methyl 9-octadecenoate, 9-octadecenoic acid, methyl ester, 9-octadecenoic acid, methyl ester, e, 9-octadecenoic acid, methyl ester, 9e, methyl e-octadec-9-enoate | |
| methyl (E)-octadec-9-enoate |
Specifications
| 1937-62-8 | |
| 220°C | |
| MFCD00066521 | |
| methyl elaidate, elaidic acid methyl ester, methyl octadec-9-enoate, methyl trans-9-octadecenoate, e-methyl octadec-9-enoate, methyl 9-octadecenoate, 9-octadecenoic acid, methyl ester, 9-octadecenoic acid, methyl ester, e, 9-octadecenoic acid, methyl ester, 9e, methyl e-octadec-9-enoate | |
| CCCCCCCCC=CCCCCCCCC(=O)OC | |
| 296.495 | |
| 296.50 | |
| Liquid |
| Yellow | |
| C19H36O2 | |
| 1 mL | |
| QYDYPVFESGNLHU-ZHACJKMWSA-N | |
| methyl (E)-octadec-9-enoate | |
| 5280590 | |
| ≥98.0% (GC) | |
| Methyl trans-9-Octadecenoate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes
Recommended Storage : Refrigerator