missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-9-Octadecenoate 98.0+%, TCI America™

Catalog No. O02261ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02261ML 1 mL
O02265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02261ML Supplier TCI America Supplier No. O02261ML

Chemical Identifiers

CAS 1937-62-8
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00066521
InChI Key QYDYPVFESGNLHU-ZHACJKMWSA-N
Synonym methyl elaidate, elaidic acid methyl ester, methyl octadec-9-enoate, methyl trans-9-octadecenoate, e-methyl octadec-9-enoate, methyl 9-octadecenoate, 9-octadecenoic acid, methyl ester, 9-octadecenoic acid, methyl ester, e, 9-octadecenoic acid, methyl ester, 9e, methyl e-octadec-9-enoate
PubChem CID 5280590
IUPAC Name methyl (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 220°C
Quantity 1 mL
Formula Weight 296.50
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-9-Octadecenoate
EINECSNumber (2)-0798
TSCA Yes
Recommended Storage Refrigerator