missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-4-Aminocyclohexanecarboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. M19861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19861G 1 g
M19865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19861G Supplier TCI America Supplier No. M19861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Physical Form: Crystalline Powder
  • Purity: ≥98.0% (N,T)
  • Molecular Formula: C8H16ClNO2
  • Molecular Weight: 193.67 g/mol
  • Storage Temperature: Room Temperature (Recommended in a cool and dark place, <15°C)
  • Application: Laboratory reagent
  • NMR: Confirmed to structure

Chemical Identifiers

CAS 61367-07-5
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
MDL Number MFCD08274538
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym methyl trans-4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl cis-4-aminocyclohexanecarboxylate hydrochloride, cis-methyl 4-aminocyclohexanecarboxylate hydrochloride, methyl 4-aminocyclohexane-1-carboxylate hydrochloride, trans-4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride, cyclohexanecarboxylic acid, 4-amino-, methyl ester, hydrochloride, trans-4-amino-cyclohexylcarboxylic acid methyl ester hydrochloride, trans-methyl 4-aminocyclohexanecarboxylate hydrochloride, 4-amino-cyclohexanecarboxylic acid methyl ester hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 193.67
Percent Purity ≥98.0% (N,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material Methyl trans-4-Aminocyclohexanecarboxylate Hydrochloride