missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl trans-3-Pentenoate 90.0+%, TCI America™
$136.08 - $454.83
Chemical Identifiers
| CAS | 20515-19-9 |
|---|---|
| Molecular Formula | C6H10O2 |
| Molecular Weight (g/mol) | 114.14 |
| MDL Number | MFCD00191576 |
| InChI Key | KJALUUCEMMPKAC-ONEGZZNKSA-N |
| Synonym | trans-3-Pentenoic Acid Methyl Ester |
| PubChem CID | 642274 |
| IUPAC Name | methyl (3E)-pent-3-enoate |
| SMILES | COC(=O)C\C=C\C |
Chemical Identifiers
| 20515-19-9 | |
| 114.14 | |
| KJALUUCEMMPKAC-ONEGZZNKSA-N | |
| 642274 | |
| COC(=O)C\C=C\C |
| C6H10O2 | |
| MFCD00191576 | |
| trans-3-Pentenoic Acid Methyl Ester | |
| methyl (3E)-pent-3-enoate |
Specifications
| 20515-19-9 | |
| 138°C | |
| MFCD00191576 | |
| 3272 | |
| KJALUUCEMMPKAC-ONEGZZNKSA-N | |
| methyl (3E)-pent-3-enoate | |
| 642274 | |
| ≥90.0% (GC) | |
| Methyl trans-3-Pentenoate |
| Yellow | |
| C6H10O2 | |
| 5 mL | |
| trans-3-Pentenoic Acid Methyl Ester | |
| COC(=O)C\C=C\C | |
| 114.14 | |
| 114.14 | |
| Liquid |
Safety and Handling
TSCA : No