missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-pentenoate, 95%

Catalog No. p-166901
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329720050 5 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC329720050 Supplier Thermo Scientific Chemicals Supplier No. 329720050

CAS: 15790-88-2 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15790-88-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00137611
InChI Key MBAHGFJTIVZLFB-SNAWJCMRSA-N
Synonym methyl 2-pentenoate, methyl trans-2-pentenoate, methyl 2e-pent-2-enoate, 2-pentenoic acid, methyl ester, 2-pentenoic acid, methyl ester, e, methyl 3-ethylacrylate, methyl 2-transpentenoate, methyl 2-trans-pentenoate, methyl e-2-pentenoate, trans-methyl pent-2-enoate
PubChem CID 5364718
SMILES CC\C=C\C(=O)OC

Specifications

Color Colorless
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4300 to 1.4320
Quantity 5 g
IUPAC Name methyl (2E)-pent-2-enoate
Formula Weight 114
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl trans-2-pentenoate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only