CAS RN 15790-88-2
IUPAC Name:
methyl (2E)-pent-2-enoate
Synonyms:
methyl (2E)-pent-2-enoate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
MBAHGFJTIVZLFB-SNAWJCMRSA-N
SMILES:
CC\C=C\C(=O)OC
1
–
1
of
1
results
1
Promotions Available
Methyl trans-2-pentenoate, 95%
Methyl trans-2-pentenoate, 95%
CAS: 15790-88-2 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.14 MDL Number: MFCD00137611 InChI Key: MBAHGFJTIVZLFB-SNAWJCMRSA-N Synonym: methyl 2-pentenoate,methyl trans-2-pentenoate,methyl 2e-pent-2-enoate,2-pentenoic acid, methyl ester,2-pentenoic acid, methyl ester, e,methyl 3-ethylacrylate,methyl 2-transpentenoate,methyl 2-trans-pentenoate,methyl e-2-pentenoate,trans-methyl pent-2-enoate PubChem CID: 5364718 SMILES: CC\C=C\C(=O)OC