missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-11-Octadecenoate 98.0+%, TCI America™

Catalog No. p-7133152
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2309100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2309100MG Supplier TCI America Supplier No. M2309100MG
Only null left

Chemical Identifiers

CAS 6198-58-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00064484
InChI Key PVVODBCDJBGMJL-CMDGGOBGSA-N
Synonym methyl vaccenate, trans-11-octadecenoic acid methyl ester, methyl trans-11-octadecenoate, methyl e-octadec-11-enoate, methyl 11-octadecenoate, unii-6o9mga9pp2, 11-octadecenoic acid, methyl ester, methyl 11e-octadec-11-enoate, 6o9mga9pp2, trans-vaccenic acid methyl ester
PubChem CID 5364432
IUPAC Name methyl (E)-octadec-11-enoate
SMILES CCCCCCC=CCCCCCCCCCC(=O)OC

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 154°C
Quantity 100 mg
Formula Weight 296.50
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl trans-11-Octadecenoate
EINECSNumber (2)-0798
TSCA No
Recommended Storage Freezer