missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl trans-11-Octadecenoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 10°C |
| Color | Yellow |
| Boiling Point | 154°C |
| Quantity | 100 mg |
| Formula Weight | 296.50 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl trans-11-Octadecenoate |
Safety and Handling
| EINECSNumber | (2)-0798 |
| TSCA | No |
| Recommended Storage | Freezer |