missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thioglycolate 98.0+%, TCI America™

Supplier:  TCI America T1287100G

 View more versions of this product

Catalog No. T1287100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Thioglycolate
Colorless
C3H6O2S
100 g
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
COC(=O)CS
106.14
106.14
Liquid
2365-48-2
49°C
MFCD00004873
1992
MKIJJIMOAABWGF-UHFFFAOYSA-N
methyl 2-sulfanylacetate
16907
≥98.0% (GC)

Chemical Identifiers

2365-48-2
106.14
MKIJJIMOAABWGF-UHFFFAOYSA-N
16907
COC(=O)CS
C3H6O2S
MFCD00004873
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
methyl 2-sulfanylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1358

RTECSNumber : AI7350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions