missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thioglycolate 98.0+%, TCI America™

Catalog No. p-7136698
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T128725G 25 g
T1287100G 100 g
T1287500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T128725G Supplier TCI America Supplier No. T128725G
Only null left

Chemical Identifiers

CAS 2365-48-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004873
InChI Key MKIJJIMOAABWGF-UHFFFAOYSA-N
Synonym methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
PubChem CID 16907
IUPAC Name methyl 2-sulfanylacetate
SMILES COC(=O)CS

Specifications

Color Colorless
Boiling Point 49°C
Quantity 25 g
UN Number 1992
Formula Weight 106.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Thioglycolate
EINECSNumber (2)-1358
RTECSNumber AI7350000
TSCA Yes