missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiocyanate, 99%

Catalog No. AAH27879AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27879AA 5 mL
AAH27879AC 25 mL
AAH27879AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH27879AA Supplier Thermo Scientific Chemicals Supplier No. H27879AA
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 556-64-9 | C2H3NS | 73.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 556-64-9
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.113
MDL Number MFCD00001830
InChI Key VYHVQEYOFIYNJP-UHFFFAOYSA-N
Synonym methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn
PubChem CID 11168
ChEBI CHEBI:61112
IUPAC Name methyl thiocyanate
SMILES CSC#N

Specifications

Melting Point -5°C
Density 1.068
Boiling Point 131°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.468
Quantity 5 mL
UN Number UN2929
Sensitivity Moisture sensitive
Formula Weight 73.11
Percent Purity 99%
Chemical Name or Material Methyl thiocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319
Hazard Statement GHS H Statement
H300-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 209-134-6
RTECSNumber XL1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only