missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thieno[3,2-b]thiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. M30011G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30011G 1 g
M30015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30011G Supplier TCI America Supplier No. M30011G

Chemical Identifiers

CAS 98800-10-3
Molecular Formula C8H6O2S2
Molecular Weight (g/mol) 198.254
MDL Number MFCD01934498
InChI Key VCLNJFSCOKHCEJ-UHFFFAOYSA-N
Synonym Thieno[3,2-b]thiophene-2-carboxylic Acid Methyl Ester
PubChem CID 2739788
IUPAC Name methyl thieno[3,2-b]thiophene-5-carboxylate
SMILES COC(=O)C1=CC2=C(S1)C=CS2

Specifications

Melting Point 98°C
Color Yellow
Quantity 1 g
Formula Weight 198.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Thieno[3,2-b]thiophene-2-carboxylate