missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiazole-2-carboxylate, 95%

Catalog No. AAH6349606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6349606 5 g
AAH6349603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6349606 Supplier Thermo Scientific Chemicals Supplier No. H6349606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55842-56-3 | C5H5NO2S | 143.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55842-56-3
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD18075580
InChI Key YRMZUOPMHMPNPG-UHFFFAOYSA-N
PubChem CID 7567660
IUPAC Name methyl 1,3-thiazole-2-carboxylate
SMILES COC(=O)C1=NC=CS1

Specifications

Quantity 5 g
Synonym methyl thiazole-2-carboxylate, methyl 2-thiazolecarboxylate, 2-thiazolecarboxylic acid methyl ester, methyl2-thiazolecarboxylate
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl thiazole-2-carboxylate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only