CAS RN 55842-56-3
IUPAC Name:
methyl 1,3-thiazole-2-carboxylate
Synonyms:
methyl 1,3-thiazole-2-carboxylate
Molecular Weight (g/mol):
143.16
Molecular Formula:
C5H5NO2S
InChi Key:
YRMZUOPMHMPNPG-UHFFFAOYSA-N
SMILES:
COC(=O)C1=NC=CS1
1
–
1
of
1
results
1
Promotions Available
Methyl thiazole-2-carboxylate, 95%
Methyl thiazole-2-carboxylate, 95%
CAS: 55842-56-3 Molecular Formula: C5H5NO2S Molecular Weight (g/mol): 143.16 MDL Number: MFCD18075580 InChI Key: YRMZUOPMHMPNPG-UHFFFAOYSA-N PubChem CID: 7567660 IUPAC Name: methyl 1,3-thiazole-2-carboxylate SMILES: COC(=O)C1=NC=CS1