missing translation for 'onlineSavingsMsg'
Learn More

Methyl Tetrolate 97.0+%, TCI America™

Catalog No. p-7136577
Change view
Click to view available options
Quantity:
1 mL
25 mL
5 mL
Catalog No. Quantity
T10531ML 1 mL
T10535ML 5 mL
T105325ML 25 mL
3 options

Catalog No. T10531ML

Supplier: TCI America T10531ML

Only null left

Chemical Identifiers

CAS 23326-27-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00009273
InChI Key UJQCANQILFWSDJ-UHFFFAOYSA-N
PubChem CID 141019
IUPAC Name methyl but-2-ynoate
SMILES CC#CC(=O)OC

Specifications

Color Yellow
Boiling Point 148°C
Quantity 1 mL
UN Number 3272
Synonym methyl 2-butynoate, methyl tetrolate, 2-butynoic acid, methyl ester, 2-butynoic acid methyl ester, tetrolic acid methyl ester, but-2-ynoic acid methyl ester, methyl2-butynoate, methyl 2-butynoate ?, pubchem12783, acmc-1ceil
Formula Weight 98.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Tetrolate