missing translation for 'onlineSavingsMsg'
Learn More

Methyl tetracosanoate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187387
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013675 250 mg
111013674 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013675 Supplier Merck Emd Millipore Supplier No. 87115250MG
Only null left

Specifications

CAS 2442-49-1
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)22 COOCH3
MDL Number MFCD00009351
Melting Point 58°C to 60°C
Boiling Point 232°C (4 mmHg)
Synonym Lignoceric acid methyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 382.66
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less