missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl t-Butyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%, Honeywell™ Burdick & Jackson
Supplier: Honeywell-Burdick and Jackson 2424
Specifications
| Methyl t-Butyl Ether | |
| 1634-04-4 | |
| 100.0000% | |
| 55.2°C | |
| −27°C | |
| C5H12O | |
| 1730942 | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 2-methoxy-2-methylpropane | |
| 15413 | |
| 88.14 | |
| HPLC |
| B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0% | |
| 0.5 AU max. at 225 nm, 0.1 AU max. at 250 nm | |
| 0.74 g/cm3 | |
| 4 x 4 L | |
| Glass Bottle | |
| UN2398 | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| CC(C)(C)OC | |
| 88.15 | |
| CHEBI:27642 | |
| >99% |
Chemical Identifiers
| 1634-04-4 | |
| 88.15 | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| CHEBI:27642 | |
| CC(C)(C)OC |
| C5H12O | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 15413 | |
| 2-methoxy-2-methylpropane |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methyl tert-butyl ether