missing translation for 'onlineSavingsMsg'
Learn More

Methyl t-Butyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%, Honeywell™ Burdick & Jackson

Catalog No. p-7111771
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001655 6 x 1 L Glass Bottle
6001654 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001655 Supplier Honeywell Chemicals Supplier No. 2421L
Only null left

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Absorbance 0.5 AU max. at 225 nm, 0.1 AU max. at 250 nm
CAS Max % 100.0000%
Density 0.74 g/cm3
Boiling Point 55.2°C
Quantity 6 x 1 L
Flash Point −27°C
Packaging Glass Bottle
UN Number UN2398
Beilstein 1730942
Formula Weight 88.14
Percent Purity >99%
Grade HPLC
Chemical Name or Material Methyl t-Butyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methyl tert-butyl ether