missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.9%, for spectroscopy, Thermo Scientific Chemicals

Catalog No. p-216839
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167851000 100 mL Glass Bottle
AC167850010 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167851000 Supplier Thermo Scientific Chemicals Supplier No. 167851000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Absorbance 0.01 max. at 360nm, 0.05 max. at 320nm, 0.1 max. at 300nm, 0.2 max. at 290nm, 0.3 max. at 280nm, 0.5 max. at 270nm
Decomposition Information >190°C
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 100 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.005% max.
Assay Percent Range 99.9%
Specific Gravity 1.1
Linear Formula (CH3)2SO
Vapor Pressure Vapor Pressure: 0.55mbar at 20°C
Viscosity 1.987 mPa.s (25°C)
Water 0.05% max.
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity 99.9%
Grade Spectroscopy
Chemical Name or Material Methyl sulfoxide, 99.9%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA TSCA

RUO – Research Use Only