missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC414885000
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
EcoSafPak™ Plastic Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414885000 500 mL EcoSafPak™ Plastic Bottle
AC414880010 1 L Plastic Bottle
AC414880025 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414885000 Supplier Thermo Scientific Chemicals Supplier No. 414885000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
CAS Min % 99.9
CAS Max % 100.0
Decomposition Information >190°C
Density 1.1000g/mL
Quantity 500 mL
Flash Point 87°C
Packaging EcoSafPak™ Plastic Bottle
Physical Form Liquid
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.01% max.
Assay Percent Range 99.9+%
Specific Gravity 1.1
Titratable Acid 0.001 meq/g max.
Vapor Pressure Vapor Pressure: 0.55mbar at 20°C
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Water 0.1% max. (Coulometric)
Beilstein 01, IV, 1277
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Boiling Point 189.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Methyl sulfoxide
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA TSCA

RUO – Research Use Only