missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals

Catalog No. ac127790025
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
100 mL
2.5 L
25 L
5 L
Packaging:
Glass Bottle
Plastic Drum
Catalog No. Quantity Packaging
AC127790025 2.5 L Glass Bottle
AC127791000 100 mL Glass Bottle
AC127790010 1 L Glass Bottle
AC127790050 5 L Plastic Drum
AC127790250 25 L Plastic Drum
5 options

Catalog No. AC127790025

Supplier: Thermo Scientific Chemicals 127790025

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 2.5 L
Flash Point 87°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.7%
Specific Gravity 1.1
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Water 0.2% max.
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity 99.7%
Grade Pure
Chemical Name or Material Methyl sulfoxide, 99.7%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA TSCA

RUO – Research Use Only