missing translation for 'onlineSavingsMsg'
Learn More

Methyl succinyl chloride, 97%

CAS: 1490-25-1 | C5H7ClO3 | 150.56 g/mol

$233.14 - $233.14

Chemical Identifiers

CAS 1490-25-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000750
InChI Key SRXOJMOGPYFZKC-UHFFFAOYSA-N
Synonym methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
PubChem CID 73888
IUPAC Name methyl 4-chloro-4-oxobutanoate
SMILES COC(=O)CCC(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108400250
SDS View Documents
Thermo Scientific Chemicals
108400250
25 g Glass bottle
Each for $233.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1490-25-1
150.56
SRXOJMOGPYFZKC-UHFFFAOYSA-N
73888
COC(=O)CCC(=O)Cl
C5H7ClO3
MFCD00000750
methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
methyl 4-chloro-4-oxobutanoate

Specifications

1490-25-1
58°C to 65°C (3.0 mmHg)
96% min. (GC)
C5H7ClO3
CH3O2CCH2CH2COCl
25 g
1.232
SRXOJMOGPYFZKC-UHFFFAOYSA-N
methyl 4-chloro-4-oxobutanoate
73888
97%
1.2320g/mL
73°C
Glass bottle
1.4402
MFCD00000750
02, II, 553
methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
COC(=O)CCC(=O)Cl
150.56
150.56
Methyl succinyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-077-

SDS
Documents

Documents

RUO – Research Use Only