missing translation for 'onlineSavingsMsg'
Learn More

Methyl Stearate 97.0+%, TCI America™

$32.28 - $58.85

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.511
MDL Number MFCD00009005
InChI Key HPEUJPJOZXNMSJ-UHFFFAOYSA-N
Synonym methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
PubChem CID 8201
ChEBI CHEBI:69188
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S008025G
View Documents
TCI America
S008025G
25 g
Each for $32.28
Only null left
 
S0080500G
View Documents
TCI America
S0080500G
500 g
Each for $58.85
Only null left
 
Description

Description

Specifications

Chemical Identifiers

112-61-8
298.511
HPEUJPJOZXNMSJ-UHFFFAOYSA-N
8201
methyl octadecanoate
C19H38O2
MFCD00009005
methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
CHEBI:69188
CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

112-61-8
White
C19H38O2
25 g
HPEUJPJOZXNMSJ-UHFFFAOYSA-N
methyl octadecanoate
8201
298.51
Crystalline Lumps
40°C
182°C
MFCD00009005
methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
CCCCCCCCCCCCCCCCCC(=O)OC
298.511
CHEBI:69188
≥97.0% (GC)
Methyl Stearate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : WI4460000

TSCA : Yes

SDS
Documents

Documents