missing translation for 'onlineSavingsMsg'
Learn More

Methyl Stearate 97.0+%, TCI America™

Catalog No. S0080500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0080500G 500 g
S008025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0080500G Supplier TCI America Supplier No. S0080500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.511
MDL Number MFCD00009005
InChI Key HPEUJPJOZXNMSJ-UHFFFAOYSA-N
Synonym methyl stearate, octadecanoic acid, methyl ester, stearic acid methyl ester, metholene 2218, kemester 9718, methyl n-octadecanoate, stearic acid, methyl ester, kemester 9018, emery 2218, n-octadecanoic acid methyl ester
PubChem CID 8201
ChEBI CHEBI:69188
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 40°C
Color White
Boiling Point 182°C
Quantity 500 g
Formula Weight 298.51
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Stearate
EINECSNumber (2)-0798
RTECSNumber WI4460000
TSCA Yes