missing translation for 'onlineSavingsMsg'
Learn More

Methyl stearate, EMD-Millipore

Catalog No. M1096020005
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096020005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096020005 Supplier Technidata Supplier No. 1.09602.0005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fatty acid methyl esters

Chemical Identifiers

CAS 112-61-8
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.511
InChI Key HPEUJPJOZXNMSJ-UHFFFAOYSA-N
Synonym Octadecanoic acid methyl ester
PubChem CID 8201
ChEBI CHEBI:69188
IUPAC Name methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Packaging Glass vials
Density Density: 0.85g/cm3 at 40°C
Assay Percent Range ≥99.0%
Quantity 5 g
Formula Weight 298.5
Chemical Name or Material Methyl stearate
Recommended Storage Store without restrictions