missing translation for 'onlineSavingsMsg'
Learn More

Methyl stearate, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012495
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012495 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012495 Supplier Merck Emd Millipore Supplier No. 857691G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl stearate is a solid, saturated fatty ester, which can be prepared from methyl oleate via catalytic hydrogenation.

Specifications

CAS 112-61-8
Physical Form Neat
Linear Formula CH3(CH2)16 CO2CH3
MDL Number MFCD00009005
Melting Point 37°C to 41°C
Boiling Point 181°C to 182°C (4 mmHg)
Synonym Methyl octadecanoate; Stearic acid methyl ester
Quantity 1 g
Formula Weight 298.50
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less